Learn More
Phenylacetyl chloride, 98%, Thermo Scientific Chemicals
44.22 € - 236.18 €
Chemical Identifiers
CAS | 103-80-0 |
---|---|
Molecular Formula | C8H7ClO |
Molecular Weight (g/mol) | 154.59 |
MDL Number | MFCD00000729 |
InChI Key | VMZCDNSFRSVYKQ-UHFFFAOYSA-N |
Synonym | phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl |
PubChem CID | 7679 |
IUPAC Name | 2-phenylacetyl chloride |
SMILES | ClC(=O)CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484030
|
Thermo Scientific Alfa Aesar
B24987.22 |
100 g |
44.22 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494030
|
Thermo Scientific Alfa Aesar
B24987.36 |
500 g |
137.69 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431567
|
Thermo Scientific Alfa Aesar
B24987.0B |
1000 g |
236.18 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phenylacetyl chloride is used in the preparation of 1,2-diphenyl-ethanone by reaction with iodobenzene using tetrabutylammonium tetrafluoroborate as a phase transfer catalyst. It is widely used in the chemical industry for manufacturing dyes and pharmaceutical products. It plays an important role in the preparation of penicillin. Further, it is used to prepare esters for flavorings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenylacetyl chloride is used in the preparation of 1,2-diphenyl-ethanone by reaction with iodobenzene using tetrabutylammonium tetrafluoroborate as a phase transfer catalyst. It is widely used in the chemical industry for manufacturing dyes and pharmaceutical products. It plays an important role in the preparation of penicillin. Further, it is used to prepare esters for flavorings.
Solubility
Miscible with alcohol and ether.
Notes
Moisture sensitive. Store in a cool place.Incompatible with amines, most common metals, strong bases and strong oxidizing agents.
Chemical Identifiers
103-80-0 | |
154.59 | |
VMZCDNSFRSVYKQ-UHFFFAOYSA-N | |
7679 | |
ClC(=O)CC1=CC=CC=C1 |
C8H7ClO | |
MFCD00000729 | |
phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl | |
2-phenylacetyl chloride |
Specifications
103-80-0 | |
225°C | |
C8H7ClO | |
MFCD00000729 | |
742254 | |
phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl | |
VMZCDNSFRSVYKQ-UHFFFAOYSA-N | |
2-phenylacetyl chloride | |
7679 | |
98% |
1.168 | |
102°C (215°F) | |
1.5325 | |
UN2577 | |
Moisture sensitive | |
Miscible with alcohol and ether. | |
ClC(=O)CC1=CC=CC=C1 | |
154.59 | |
154.6 | |
Phenylacetyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H500
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHENYLACETYL CHLORIDE
missing translation for 'einecsNumber' : 203-146-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.