Learn More
Phenylacetyl chloride, 98%, Thermo Scientific Chemicals
Description
Phenylacetyl chloride is used in the preparation of 1,2-diphenyl-ethanone by reaction with iodobenzene using tetrabutylammonium tetrafluoroborate as a phase transfer catalyst. It is widely used in the chemical industry for manufacturing dyes and pharmaceutical products. It plays an important role in the preparation of penicillin. Further, it is used to prepare esters for flavorings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phenylacetyl chloride is used in the preparation of 1,2-diphenyl-ethanone by reaction with iodobenzene using tetrabutylammonium tetrafluoroborate as a phase transfer catalyst. It is widely used in the chemical industry for manufacturing dyes and pharmaceutical products. It plays an important role in the preparation of penicillin. Further, it is used to prepare esters for flavorings.
Solubility
Miscible with alcohol and ether.
Notes
Moisture sensitive. Store in a cool place.Incompatible with amines, most common metals, strong bases and strong oxidizing agents.
Specifications
Specifications
Boiling Point | 225°C |
Flash Point | 102°C (215°F) |
Quantity | 1000 g |
UN Number | UN2577 |
Beilstein | 742254 |
Formula Weight | 154.6 |
Percent Purity | 98% |
Chemical Name or Material | Phenylacetyl chloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.