missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Solution, Jenway™
Protection and care for pH electrodes
56.80 € - 60.45 €
Product Code | Brand | pH | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH | Price | Quantity & Availability | |||||
11470579
|
Jenway
90238-53 |
2 |
56.80 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460579
|
Jenway
16156-78 |
9.22 |
60.45 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Storage solution for pH electrodes
- 500mL bottle
Specifications
500 mL |