missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH Buffer Solution, Jenway™
Click to view available options
pH:
2
9.22
Description
- Storage solution for pH electrodes
- 500mL bottle
Specifications
Specifications
pH | 9.22 |
Quantity | 500 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'