missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periodic Acid, puriss p.a, ACS, ≥99.0% (RT), Honeywell™ Fluka™
puriss. p.a., ACS reagent, for oxidimetric titration, crystallized,≥99.0% (RT)
106.65 € - 1235.40 €
Chemical Identifiers
CAS | 10450-60-9 |
---|---|
Molecular Formula | H5IO6 |
Molecular Weight (g/mol) | 227.94 |
MDL Number | MFCD00011344 |
InChI Key | TWLXDPFBEPBAQB-UHFFFAOYSA-N |
Synonym | orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst |
PubChem CID | 25289 |
ChEBI | CHEBI:29150 |
SMILES | O[I](O)(O)(O)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15657060
|
Honeywell-Fluka
77310-25G |
25 g | Glass bottle | ||||||
15647060
|
Honeywell-Fluka
77310-100G |
100 g | Glass bottle | ||||||
15667060
|
Honeywell Research Chemicals
77312-100G |
100 g | Glass bottle | ||||||
15677060
|
Honeywell Research Chemicals
77312-500G |
500 g | Glass bottle | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10450-60-9 | |
227.94 | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
25289 | |
O[I](O)(O)(O)(O)=O |
H5IO6 | |
MFCD00011344 | |
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
CHEBI:29150 |
Specifications
White | |
H5IO6 | |
MFCD00011344 | |
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
pentahydroxy(oxo)-$l^{7}-iodane | |
25289 | |
227.94g/mol | |
Periodic acid |
10450-60-9 | |
Solid | |
UN3085 | |
Soluble in water | |
O[I](O)(O)(O)(O)=O | |
227.94 | |
CHEBI:29150 | |
≥99.0% (RT) |
Safety and Handling
P220-P280-P305 + P351 + P338-P310
missing translation for 'dotInformation' : 8(5.1) / PGII
missing translation for 'rtecsNumber' : 233-937-0