missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, puriss p.a, ACS, ≥99.0% (RT), Honeywell™ Fluka™

Product Code. 15667060
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

puriss. p.a., ACS reagent, for oxidimetric titration, crystallized,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
SMILES O[I](O)(O)(O)(O)=O

Specifications

Color White
Packaging Glass bottle
Quantity 100 g
Physical Form Solid
UN Number UN3085
Solubility Information Soluble in water
IUPAC Name tetrahydroxyiodous acid
Formula Weight 227.94g/mol
Percent Purity ≥99.0% (RT)
Chemical Name or Material Periodic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'