Learn More
Perfluoro-compound FC-40(TM), Thermo Scientific Chemicals
40.80 € - 396.98 €
Chemical Identifiers
CAS | 51142-49-5 |
---|---|
Molecular Formula | C21F48N2 |
Molecular Weight (g/mol) | 1192.168 |
MDL Number | MFCD01632188 |
InChI Key | QDOIZVITZUBGOQ-UHFFFAOYSA-N |
Synonym | fluorinert fc-40, perfluoro-compound fc-40, bis nonafluorobutyl trifluoromethyl amine; perfluorotributylamine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine,1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine; 1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl trifluoromethyl amine; perfluorotributylamine |
PubChem CID | 2723673 |
IUPAC Name | 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)-N-(trifluoromethyl)butan-1-amine |
SMILES | C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F.C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(F)(F)F)(F)F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10163360
|
Thermo Scientific Acros
123760100 |
10 g | Glass bottle |
40.80 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10081150
|
Thermo Scientific Acros
123761000 |
100 g | Glass bottle |
216.08 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173360
|
Thermo Scientific Acros
123762500 |
250 g | Glass bottle |
396.98 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51142-49-5 | |
1192.168 | |
QDOIZVITZUBGOQ-UHFFFAOYSA-N | |
2723673 | |
C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F.C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(F)(F)F)(F)F)(F)F |
C21F48N2 | |
MFCD01632188 | |
fluorinert fc-40, perfluoro-compound fc-40, bis nonafluorobutyl trifluoromethyl amine; perfluorotributylamine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine,1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine; 1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl trifluoromethyl amine; perfluorotributylamine | |
1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)-N-(trifluoromethyl)butan-1-amine |
Specifications
51142-49-5 | |
1.8700g/mL | |
Authentic | |
1.29 | |
1.87 | |
QDOIZVITZUBGOQ-UHFFFAOYSA-N | |
1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)-N-(trifluoromethyl)butan-1-amine | |
2723673 | |
650 | |
Perfluoro-compound FC-40™ |
Colorless | |
158.0°C to 173.0°C | |
C21F48N2 | |
MFCD01632188 | |
fluorinert fc-40, perfluoro-compound fc-40, bis nonafluorobutyl trifluoromethyl amine; perfluorotributylamine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine,1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine; 1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl trifluoromethyl amine; perfluorotributylamine | |
C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F.C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(F)(F)F)(F)F)(F)F | |
1192.168 | |
1.6-2.8 mm2/s (25°C) | |
Liquid |
Safety and Handling
GHS H Statement:
May cause long lasting harmful effects to aquatic life.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- Perfluoro-compound FC-40]TS
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
RUO – Research Use Only