missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-compound FC-40(TM), Thermo Scientific Chemicals

Product Code. 10081150
Click to view available options
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51142-49-5
Molecular Formula C21F48N2
Molecular Weight (g/mol) 1192.168
MDL Number MFCD01632188
InChI Key QDOIZVITZUBGOQ-UHFFFAOYSA-N
Synonym fluorinert fc-40, perfluoro-compound fc-40, bis nonafluorobutyl trifluoromethyl amine; perfluorotributylamine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine,1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine; 1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl trifluoromethyl amine; perfluorotributylamine
PubChem CID 2723673
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)-N-(trifluoromethyl)butan-1-amine
SMILES C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F.C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Density 1.8700g/mL
Boiling Point 158°C to 173°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Viscosity 1.6-2.8cSt (25°C)
Formula Weight 650
Physical Form Liquid
Chemical Name or Material Perfluoro-compound FC-40™
Product Identifier
  • Perfluoro-compound FC-40]TS
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'