missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane, ≥99%,Spectrophotometric Grade, Honeywell™ Riedel-de-Haën™
109.65 € - 216.34 €
Chemical Identifiers
CAS | 109-66-0 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009498 |
InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
PubChem CID | 8003 |
ChEBI | CHEBI:37830 |
IUPAC Name | pentane |
SMILES | CCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15601470
|
Honeywell Riedel-de Haën
154954-1L |
1 L | Glass Bottle | ||||||
15611470
|
Honeywell Riedel-de Haën
154954-2L |
2 L | Glass Bottle |
216.34 €
2 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
109-66-0 | |
72.15 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
8003 | |
pentane |
C5H12 | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CHEBI:37830 | |
CCCCC |
Specifications
-131°C | |
0.626g/mL at 25°C | |
0.083 | |
500°F | |
C5H12 | |
MFCD00009498 | |
UN1265 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CCCCC | |
72.15 | |
CHEBI:37830 | |
≥99% | |
Pentane |
109-66-0 | |
36°C to 37°C | |
−49°C (−56.2°F) | |
1.356 | |
CH3(CH2)3CH3 | |
26.98psi (55°C), 8.4psi (20°C) | |
969132 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15g/mol | |
Spectrophotometric |
Safety and Handling
P210-P301 + P310-P331-P370 + P378-P403 + P235
H225-H304-H336-H411
missing translation for 'dotInformation' : Class 3 - PG 2 - Pentanes
missing translation for 'einecsNumber' : 203-692-4
missing translation for 'rtecsNumber' : RZ9450000