missing translation for 'onlineSavingsMsg'
Learn More

Pentane, ≥99%,Spectrophotometric Grade, Honeywell™ Riedel-de-Haën™

Product Code. 15601470
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -131°C
Density 0.626g/mL at 25°C
Boiling Point 36°C to 37°C
Quantity 1 L
Explosion Limit 0.083
Flash Point −49°C (−56.2°F)
Ignition Point 500°F
Packaging Glass Bottle
Refractive Index 1.356
Linear Formula CH3(CH2)3CH3
Vapor Pressure 26.98psi (55°C), 8.4psi (20°C)
UN Number UN1265
Beilstein 969132
Formula Weight 72.15g/mol
Percent Purity ≥99%
Grade Spectrophotometric
Chemical Name or Material Pentane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'