Learn More
Palladium(II) nitrate hydrate, Thermo Scientific Chemicals
152.76 € - 3255.20 €
Chemical Identifiers
CAS | 207596-32-5, 10102-05-3 |
---|---|
Molecular Formula | N2O6Pd |
Molecular Weight (g/mol) | 230.43 |
MDL Number | MFCD00149818 |
InChI Key | GPNDARIEYHPYAY-UHFFFAOYSA-N |
Synonym | palladium ii nitrate hydrate, palladium nitrate hydrate, palladium 2+ hydrate dinitrate, acmc-1bnae, palladous nitrate hydrate, palladium dinitrate hydrate, 2no3.pd.h2o, ksc492s0r, palladium ii nitrate,hydrate, palladium,diaquabis nitrato-o-9ci |
PubChem CID | 16211503 |
SMILES | [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10279243
|
Thermo Scientific Acros
389910010 |
1 g | Glass bottle |
152.76 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10239293
|
Thermo Scientific Acros
389910050 |
5 g | Glass bottle |
763.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11994361
|
Thermo Scientific Acros
389910250 |
25 g | Glass bottle |
3255.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207596-32-5, 10102-05-3 | |
230.43 | |
GPNDARIEYHPYAY-UHFFFAOYSA-N | |
16211503 |
N2O6Pd | |
MFCD00149818 | |
palladium ii nitrate hydrate, palladium nitrate hydrate, palladium 2+ hydrate dinitrate, acmc-1bnae, palladous nitrate hydrate, palladium dinitrate hydrate, 2no3.pd.h2o, ksc492s0r, palladium ii nitrate,hydrate, palladium,diaquabis nitrato-o-9ci | |
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
207596-32-5, 10102-05-3 | |
Crystals | |
Pd(NO3)2·xH2O | |
palladium ii nitrate hydrate, palladium nitrate hydrate, palladium 2+ hydrate dinitrate, acmc-1bnae, palladous nitrate hydrate, palladium dinitrate hydrate, 2no3.pd.h2o, ksc492s0r, palladium ii nitrate,hydrate, palladium,diaquabis nitrato-o-9ci | |
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
230.43 | |
230.43 |
Black-Brown | |
N2O6Pd | |
MFCD00149818 | |
GPNDARIEYHPYAY-UHFFFAOYSA-N | |
palladium(2+) dinitrate | |
16211503 | |
Palladium(II) nitrate hydrate |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
- Palladium(II) nitrate hydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only