missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium(II) nitrate hydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Black-Brown |
Physical Form | Crystals |
Quantity | 25 g |
Linear Formula | Pd(NO3)2·xH2O |
IUPAC Name | palladium(2+) dinitrate |
Formula Weight | 230.43 |
Packaging | Glass bottle |
Chemical Name or Material | Palladium(II) nitrate hydrate |
Safety and Handling
Product Identifier
- Palladium(II) nitrate hydrate
- Danger
- Acute toxicity Category 4
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'