missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 0.12%, Ruthenium, 0.12%; on 3mm alumina tablets, Thermo Scientific™
90.25 € - 325.00 €
Chemical Identifiers
CAS | 7440-05-3 |
---|---|
Molecular Formula | Pd |
Molecular Weight (g/mol) | 106.42 |
MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
Synonym | black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon |
PubChem CID | 23938 |
ChEBI | CHEBI:33363 |
IUPAC Name | palladium |
SMILES | [Pd] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Reduction of nitrogen oxides to N2 with H2 in the presence of CO and CO2
Chemical Identifiers
7440-05-3 | |
106.42 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
23938 | |
palladium |
Pd | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
CHEBI:33363 | |
[Pd] |
Specifications
7440-05-3 | |
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
palladium | |
23938 | |
0.12%, Ruthenium, 0.12%; on 3mm alumin.a tablets |
Pd | |
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
[Pd] | |
106.42 | |
CHEBI:33363 | |
Palladium |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures