missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 0.12%, Ruthenium, 0.12%; on 3mm alumina tablets, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Reduction of nitrogen oxides to N2 with H2 in the presence of CO and CO2
Specifications
Specifications
Quantity | 50 g |
Concentration or Composition (by Analyte or Components) | 0.12%, Ruthenium, 0.12%; on 3mm alumin.a tablets |
Chemical Name or Material | Palladium |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'