missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P-Xylene, ReagentPlus™, 99%, Honeywell™
Reagent Grade, 99%
36.13 € - 947.53 €
Chemical Identifiers
CAS | 106-42-3 |
---|---|
Molecular Formula | C8H10 |
Molecular Weight (g/mol) | 106.17 |
MDL Number | MFCD00008556 |
InChI Key | URLKBWYHVLBVBO-UHFFFAOYSA-N |
Synonym | p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene |
PubChem CID | 7809 |
ChEBI | CHEBI:27417 |
IUPAC Name | 1,4-xylene |
SMILES | CC1=CC=C(C)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15631330
|
Honeywell Research Chemicals
134449-500ML |
500 mL | Glass Bottle | ||||||
15611330
|
Honeywell Research Chemicals
134449-2.5L |
2.5 L | Glass Bottle | ||||||
15621330
|
Honeywell Research Chemicals
134449-20L |
20 L | Stainless Steel Can | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
106-42-3 | |
106.17 | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
7809 | |
1,4-xylene |
C8H10 | |
MFCD00008556 | |
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene | |
CHEBI:27417 | |
CC1=CC=C(C)C=C1 |
Specifications
12°C | |
Colorless | |
137°C to 139°C | |
25°C (77°F) | |
Liquid | |
C8H10 | |
MFCD00008556 | |
UN1307 | |
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene | |
URLKBWYHVLBVBO-UHFFFAOYSA-N | |
1,4-xylene | |
7809 | |
106.17g/mol | |
ReagentPlus™ |
106-42-3 | |
0.861g/mL at 20°C | |
0.07 | |
984°F | |
n20/D 1.495(lit.) | |
C6H4(CH3)2 | |
9mmHg (20°C) | |
1901563 | |
Soluble in water, 0.2g/L | |
CC1=CC=C(C)C=C1 | |
106.17 | |
CHEBI:27417 | |
99% | |
P-Xylene |
Safety and Handling
P210-P261-P280-P370 + P378
H226-H312 + H332-H315
missing translation for 'dotInformation' : class 3 - PG 3 - Xylenes
missing translation for 'einecsNumber' : 203-396-5
missing translation for 'rtecsNumber' : ZE2625000