missing translation for 'onlineSavingsMsg'
Learn More

P-Xylene, ReagentPlus™, 99%, Honeywell™

Product Code. 15621330
Click to view available options
Quantity:
500 mL
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1

Specifications

Melting Point 12°C
Color Colorless
Density 0.861g/mL at 20°C
Boiling Point 137°C to 139°C
Quantity 20 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Ignition Point 984°F
Packaging Stainless Steel Can
Physical Form Liquid
Refractive Index n20/D 1.495(lit.)
Linear Formula C6H4(CH3)2
Vapor Pressure 9mmHg (20°C)
UN Number UN1307
Beilstein 1901563
Solubility Information Soluble in water, 0.2g/L
Formula Weight 106.17g/mol
Percent Purity 99%
Grade ReagentPlus™
Chemical Name or Material P-Xylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.