missing translation for 'onlineSavingsMsg'
Learn More

Osteopontin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen Osteopontin
Concentration 0.28 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16864275
View Documents
Proteintech
25715-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16854275
View Documents
Proteintech
25715-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Osteopontin is an arginine-glycine-aspartic acid (RGD)-containing glycoprotein that interacts with integrins and CD44 as major receptors. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. Proteolytic cleavage by thrombin and matrix metalloproteinases close to the integrin-binding Arg-Gly-Asp sequence modulates the function of OPN and its integrin binding properties. Thrombin-cleaved fragments of Osteopontin are overexpressed in maligt glial tumors and provide a molecular niche with survival advantage and provide a novel substrate for plasmin and cathepsin D.
Specifications

Specifications

Osteopontin
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Rat, Human
P08721, P10451, P10923
20750, 25353, 6696
Osteopontin Fusion Protein Ag22588
Primary
-20°C
SPP1
0.28 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
BNSP, Bone sialoprotein 1, BSPI, ETA 1, Nephropontin, OPN, Osteopontin, secreted phosphoprotein 1, SPP 1, SPP1, Urinary stone protein, Uropontin
SPP1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.