missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteopontin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25715-1-AP-20UL
This item is not returnable.
View return policy
Description
Osteopontin is an arginine-glycine-aspartic acid (RGD)-containing glycoprotein that interacts with integrins and CD44 as major receptors. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. Proteolytic cleavage by thrombin and matrix metalloproteinases close to the integrin-binding Arg-Gly-Asp sequence modulates the function of OPN and its integrin binding properties. Thrombin-cleaved fragments of Osteopontin are overexpressed in maligt glial tumors and provide a molecular niche with survival advantage and provide a novel substrate for plasmin and cathepsin D.Specifications
Osteopontin | |
Polyclonal | |
Unconjugated | |
SPP1 | |
BNSP, Bone sialoprotein 1, BSPI, ETA 1, Nephropontin, OPN, Osteopontin, secreted phosphoprotein 1, SPP 1, SPP1, Urinary stone protein, Uropontin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20750, 25353, 6696 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P08721, P10451, P10923 | |
SPP1 | |
Osteopontin Fusion Protein Ag22588 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction