missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Nylon Threaded Nipple

Threaded fittings provide a more permanent fluid path

Brand:  Masterflex™ 30623-25

 View more versions of this product

Product Code. 11741104

  • 26.60 € / Pack of 6
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Threaded Nipple NPT(M)
  • Use with our rigid tubing for high-pressure applications
  • Good chemical resistance
  • Rated to 200psi at 77°F (25°C)
  • Usable from -40 to +200°F (-40 to +93°C)
Specifications

Specifications

Threaded Nipple NPT(M)
Not Autoclavable
1/8 in.
1/8 in.
1/8 in.
NPT(M) Connection
Nylon
1/8 in.
Ethylene Oxide
Translucent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers