Learn More
Macherey-Nagel™ Nucleodur™ NH2-RP HPLC EC Guard Columns
Brand:  Macherey-Nagel™ 761951.20
 View more versions of this product
Product Code. 12758982
  • 161.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
2.5%
4 mm
EC Guard Column
NUCLEODUR™ NH2-RP
Acetonitrile/Water
Aminopropyl
Polar Compounds Under RP Conditions and Hydrocarbons Under NP Conditions
Spherical
L8
3 μm
100Å
2 mm
Acetonitrile/Water
Silica
Guard Column
HPLC
No