missing translation for 'onlineSavingsMsg'
Learn More

NF-L Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5C12G4, Proteintech

Mouse Monoclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen NF-L
Clone 5C12G4
Concentration 1.4 mg/mL
Applications Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16867175
View Documents
Proteintech
60189-1-IG-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16857175
View Documents
Proteintech
60189-1-IG-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is specific to NEFL.

Involved in the maintece of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220kDa, with some variation in different species.
Specifications

Specifications

NF-L
1.4 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
68 kDa neurofilament protein, NEFL, Neurofilament L, NF68, NFL, NF-L
Nefl
IgG1
Protein G
Antibody
5C12G4
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Mouse
Human, Mouse, Rat, Porcine
P02547, P07196, P08551, P19527
100521224, 18039, 4747, 83613
NF-L Fusion Protein Ag15178
Primary
-20°C
NEFL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.