missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-L Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5C12G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60189-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to NEFL.
Involved in the maintece of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220kDa, with some variation in different species.Specifications
NF-L | |
Monoclonal | |
1.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02547, P07196, P08551, P19527 | |
Nefl | |
NF-L Fusion Protein Ag15178 | |
20 μL | |
Primary | |
Human, Mouse, Rat, Porcine | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
5C12G4 | |
Unconjugated | |
NEFL | |
68 kDa neurofilament protein, NEFL, Neurofilament L, NF68, NFL, NF-L | |
Mouse | |
Protein G | |
RUO | |
100521224, 18039, 4747, 83613 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction