missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpentylamine, 98%, Thermo Scientific Chemicals

96.38 € - 348.74 €

Chemical Identifiers

CAS 25419-06-1
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00041354
InChI Key UOIWOHLIGKIYFE-UHFFFAOYSA-N
Synonym n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine
PubChem CID 117479
IUPAC Name N-methylpentan-1-amine
SMILES CCCCCNC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381737
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L04882.04
2 g
96.38 €
2g
Please to purchase this item. Need a web account? Register with us today!
11391737
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L04882.09
10 g
348.74 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25419-06-1
101.19
UOIWOHLIGKIYFE-UHFFFAOYSA-N
117479
CCCCCNC
C6H15N
MFCD00041354
n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine
N-methylpentan-1-amine

Specifications

25419-06-1
117°C to 119°C
C6H15N
MFCD00041354
1731333
n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine
CCCCCNC
101.19
101.19
N-Methylpentylamine
0.738
18°C (64°F)
1.4105
UN2733
Air Sensitive
UOIWOHLIGKIYFE-UHFFFAOYSA-N
N-methylpentan-1-amine
117479
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 246-966-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only