missing translation for 'onlineSavingsMsg'
Learn More

N-Methylpentylamine, 98%, Thermo Scientific Chemicals

Product Code. 11381737
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25419-06-1
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00041354
InChI Key UOIWOHLIGKIYFE-UHFFFAOYSA-N
Synonym n-methylpentylamine, n-methylamylamine, amylmethylamine, methyl pentyl amine, 1-pentanamine, n-methyl, methylpentylamine, pentylamine, n-methyl, methyl-pentyl-amine, n-methyl-n-pentylamine, methylamylamine
PubChem CID 117479
IUPAC Name N-methylpentan-1-amine
SMILES CCCCCNC

Specifications

Density 0.738
Boiling Point 117°C to 119°C
Flash Point 18°C (64°F)
Refractive Index 1.4105
Quantity 2 g
UN Number UN2733
Beilstein 1731333
Sensitivity Air Sensitive
Formula Weight 101.19
Percent Purity 98%
Chemical Name or Material N-Methylpentylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'