Learn More
N-Benzyl-N-ethylaniline, 98%, Thermo Scientific™
16.50 € - 34.00 €
Chemical Identifiers
CAS | 92-59-1 |
---|---|
Molecular Formula | C15H17N |
Molecular Weight (g/mol) | 211.31 |
MDL Number | MFCD00009037 |
InChI Key | HSZCJVZRHXPCIA-UHFFFAOYSA-N |
Synonym | ethylbenzylaniline, benzenemethanamine, n-ethyl-n-phenyl, n-ethyl-n-phenylbenzylamine, benzylethylphenylamine, phenylethylbenzylamine, n-ethyl-n-benzylaniline, benzylamine, n-ethyl-n-phenyl, n-ethyl-n-phenyl benzylamine, n-ethyl-n-phenylbenzenemethanamine, amine, benzyl ethyl phenyl |
PubChem CID | 7098 |
IUPAC Name | N-benzyl-N-ethylaniline |
SMILES | CCN(CC1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448717
|
Thermo Scientific Alfa Aesar
A19882.09 |
10 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11458717
|
Thermo Scientific Alfa Aesar
A19882.18 |
50 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11468717
|
Thermo Scientific Alfa Aesar
A19882.30 |
250 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
92-59-1 | |
211.31 | |
HSZCJVZRHXPCIA-UHFFFAOYSA-N | |
7098 | |
CCN(CC1=CC=CC=C1)C1=CC=CC=C1 |
C15H17N | |
MFCD00009037 | |
ethylbenzylaniline, benzenemethanamine, n-ethyl-n-phenyl, n-ethyl-n-phenylbenzylamine, benzylethylphenylamine, phenylethylbenzylamine, n-ethyl-n-benzylaniline, benzylamine, n-ethyl-n-phenyl, n-ethyl-n-phenyl benzylamine, n-ethyl-n-phenylbenzenemethanamine, amine, benzyl ethyl phenyl | |
N-benzyl-N-ethylaniline |
Specifications
92-59-1 | |
163°C to 164°C (6 mmHg) | |
C15H17N | |
MFCD00009037 | |
2099852 | |
ethylbenzylaniline, benzenemethanamine, n-ethyl-n-phenyl, n-ethyl-n-phenylbenzylamine, benzylethylphenylamine, phenylethylbenzylamine, n-ethyl-n-benzylaniline, benzylamine, n-ethyl-n-phenyl, n-ethyl-n-phenyl benzylamine, n-ethyl-n-phenylbenzenemethanamine, amine, benzyl ethyl phenyl | |
CCN(CC1=CC=CC=C1)C1=CC=CC=C1 | |
211.31 | |
211.31 | |
N-Benzyl-N-ethylaniline |
1.03 | |
>157°C (315°F) | |
1.595 | |
UN2274 | |
14,3769 | |
HSZCJVZRHXPCIA-UHFFFAOYSA-N | |
N-benzyl-N-ethylaniline | |
7098 | |
98% |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: N-ETHYL-N-BENZYLANILINE
missing translation for 'einecsNumber' : 202-169-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures