missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-N-ethylaniline, 98%, Thermo Scientific™

Product Code. 11458717
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-59-1
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.31
MDL Number MFCD00009037
InChI Key HSZCJVZRHXPCIA-UHFFFAOYSA-N
Synonym ethylbenzylaniline, benzenemethanamine, n-ethyl-n-phenyl, n-ethyl-n-phenylbenzylamine, benzylethylphenylamine, phenylethylbenzylamine, n-ethyl-n-benzylaniline, benzylamine, n-ethyl-n-phenyl, n-ethyl-n-phenyl benzylamine, n-ethyl-n-phenylbenzenemethanamine, amine, benzyl ethyl phenyl
PubChem CID 7098
IUPAC Name N-benzyl-N-ethylaniline
SMILES CCN(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.03
Boiling Point 163°C to 164°C (6 mmHg)
Flash Point >157°C (315°F)
Refractive Index 1.595
Quantity 50 g
UN Number UN2274
Beilstein 2099852
Merck Index 14,3769
Formula Weight 211.31
Percent Purity 98%
Chemical Name or Material N-Benzyl-N-ethylaniline
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'