missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RapidPURE™ Plasmid Mini Kit

RapidPURE™ Plasmid Mini Kit for use in routine molecular biology applications such as fluorescent and radioactive sequencing and cloning.

Brand:  MP Biomedicals™ 112066600

 View more versions of this product

Product Code. 15797879

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The Kit is designed to isolate up to 20 μg of plasmid DNA from 1.5 mL of an overnight bacterial culture.
  • All steps can be carried out in either a microcentrifuge or vacuum manifold.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at Room temperature (15° to 30°C)
Plasmid Prep
300 Preps
1.5 mL of Overnight Culture
Kit
Isolate up to 20 μg of plasmid DNA from 1.5 mL of an overnight bacterial culture
Plasmid isolation kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.