missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS, 1.0M buffer soln., pH 9.0, Thermo Scientific™
72.70 € - 94.00 €
Chemical Identifiers
CAS | 1132-61-2 |
---|---|
Molecular Formula | C7H15NO4S |
Molecular Weight (g/mol) | 209.26 |
MDL Number | MFCD00006183 |
InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
PubChem CID | 70807 |
ChEBI | CHEBI:44115 |
IUPAC Name | 3-morpholin-4-ylpropane-1-sulfonic acid |
SMILES | [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15472929
|
Thermo Scientific Alfa Aesar
J60097.AK |
250 mL |
72.70 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15482929
|
Thermo Scientific Alfa Aesar
J60097.AP |
500 mL | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1132-61-2 | |
209.26 | |
DVLFYONBTKHTER-UHFFFAOYSA-N | |
CHEBI:44115 | |
[O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
C7H15NO4S | |
MFCD00006183 | |
70807 | |
3-morpholin-4-ylpropane-1-sulfonic acid |
Specifications
1132-61-2 | |
1.0 M | |
Liquid | |
Odorless | |
DVLFYONBTKHTER-UHFFFAOYSA-N | |
3-morpholin-4-ylpropane-1-sulfonic acid | |
70807 | |
MOPS buffer soln. |
Colorless | |
9.0 | |
C7H15NO4S | |
MFCD00006183 | |
[O-]S(=O)(=O)CCC[NH+]1CCOCC1 | |
209.26 | |
CHEBI:44115 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures