missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 1.0M buffer soln., pH 9.0, Thermo Scientific™

Product Code. 15482929
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Color Colorless
Concentration 1.0 M
pH 9.0
Physical Form Liquid
Quantity 500 mL
Odor Odorless
Chemical Name or Material MOPS buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'