missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 4A, pellets, 3.2mm diameter, Thermo Scientific Chemicals

140.70 € - 381.90 €

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10882231
View Documents Promotion Details
Thermo Scientific Acros
436730010
1 kg Glass Bottle
140.70 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10535090
View Documents Promotion Details
Thermo Scientific Acros
436730050
5 kg Plastic drum
381.90 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
Specifications

Chemical Identifiers

70955-01-0
440.42
FJUZEZRZKVMAMD-UHFFFAOYNA-N
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
C20H25FN2O8
MFCD00131613
73906282

Specifications

Beige
< 2% (900°C, 2 hrs.)
70955-01-0
C20H25FN2O8
FJUZEZRZKVMAMD-UHFFFAOYNA-N
Molecular sieves
73906282
650 to 750g/L
> 20% (55% rH, 20°C)
Cylinders
MFCD00131613
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
440.42
Molecular sieves 4A
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only