missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 4A, pellets, 3.2mm diameter, Thermo Scientific Chemicals

Product Code. 10535090
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass Bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Color Beige
Bulk Density 650 to 750g/L
Packaging Plastic drum
Residual Water < 2% (900°C, 2 hrs.)
Water Adsorption > 20% (55% rH, 20°C)
Quantity 5 kg
Physical Form Cylinders
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
Chemical Name or Material Molecular sieves 4A

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'