missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ MN 615 Grade Round Format Cellulose Qualitative Filter Paper

Use for a wide range of general laboratory filtrations. MN 615 Grade Round Format Cellulose Qualitative Filter Paper provides medium fast filtration.

6.30 € - 20.10 €

Specifications

Format Disc
Material Cellulose
Filtration Speed Medium fast
For Use With (Application) Qualitative analysis
Grade MN 615
View More Specs

Products 7
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10473881
View Documents Promotion Details
Macherey-Nagel™
431011
110 mm
10.20 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10220891
View Documents Promotion Details
Macherey-Nagel™
431007
70 mm
6.30 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10720612
View Documents Promotion Details
Macherey-Nagel™
431015
150 mm
14.70 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10067870
View Documents Promotion Details
Macherey-Nagel™
431012
125 mm
11.10 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10322861
View Documents Promotion Details
Macherey-Nagel™
431009
90 mm
7.70 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10537711
View Documents Promotion Details
Macherey-Nagel™
431018
180 mm
20.10 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10597511
View Documents Promotion Details
Macherey-Nagel™
431005
55 mm
6.30 €
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For general purpose filtrations
  • Qualitative filter papers are manufactured from the same raw materials as the ashless grades and are particularly suited for general laboratory filtrations
  • The average ash content is about 0.1%
  • The amount of a-cellulose is about 95%.
  • Medium fast filtration
Specifications

Specifications

Disc
Medium fast
MN 615
0.16 mm
Cellulose
Qualitative analysis
100 Pack
Filter Paper
Videos
SDS
Documents

Documents

Product Certifications