missing translation for 'onlineSavingsMsg'
Learn More

Methyltrioxorhenium (VII), 98%, Thermo Scientific Chemicals

47.70 € - 628.13 €

Chemical Identifiers

CAS 70197-13-6
Molecular Formula CH3O3Re
Molecular Weight (g/mol) 249.23
MDL Number MFCD00192332
InChI Key VZSXFJPZOCRDPW-UHFFFAOYSA-N
Synonym methyltrioxorhenium vii, methyltrioxorhenium, methylrhenium trioxide, unii-883d8rdd5q, methylrhenium vii trioxide, mto, methyltrioxorhenium mi, t-4-methyltrioxorhenium, methylrhenium trioxide vii
PubChem CID 2734010
IUPAC Name carbanide;trioxorhenium
SMILES [CH3-].O=[Re](=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10081661
View Documents
Thermo Scientific Acros
346961000
100 mg Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10259253
View Documents
Thermo Scientific Acros
346965000
500 mg Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10390784
View Documents
Thermo Scientific Acros
346960025
2.5 g Glass bottle
628.13 €
2.50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

70197-13-6
249.23
VZSXFJPZOCRDPW-UHFFFAOYSA-N
2734010
[CH3-].O=[Re](=O)=O
CH3O3Re
MFCD00192332
methyltrioxorhenium vii, methyltrioxorhenium, methylrhenium trioxide, unii-883d8rdd5q, methylrhenium vii trioxide, mto, methyltrioxorhenium mi, t-4-methyltrioxorhenium, methylrhenium trioxide vii
carbanide;trioxorhenium

Specifications

111.0°C
Blue-Gray to White
100.0
97.5% min. (GC)
MFCD00192332
methyltrioxorhenium vii, methyltrioxorhenium, methylrhenium trioxide, unii-883d8rdd5q, methylrhenium vii trioxide, mto, methyltrioxorhenium mi, t-4-methyltrioxorhenium, methylrhenium trioxide vii
VZSXFJPZOCRDPW-UHFFFAOYSA-N
carbanide;trioxorhenium
2734010
98%
Methyltrioxorhenium (VII)
70197-13-6
97.5
Crystals
CH3O3Re
15, 6206
Solubility in water: very soluble in h2o
[CH3-].O=[Re](=O)=O
249.23
249.23
Authentic
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methyltrioxorhenium (VII)

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.