missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyltrioxorhenium (VII), 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 111.0°C |
Color | Blue-Gray to White |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Physical Form | Crystals |
Quantity | 500 mg |
Assay Percent Range | 97.5% min. (GC) |
Merck Index | 15, 6206 |
Solubility Information | Solubility in water: very soluble in h2o |
Formula Weight | 249.23 |
Show More |
Safety and Handling
Product Identifier
- Methyltrioxorhenium (VII)
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'