Learn More
Mercury(II) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
78.89 € - 537.00 €
Chemical Identifiers
CAS | 7487-94-7 |
---|---|
Molecular Formula | Cl2Hg |
Molecular Weight (g/mol) | 271.49 |
MDL Number | MFCD00011041 |
InChI Key | LWJROJCJINYWOX-UHFFFAOYSA-L |
Synonym | mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 |
PubChem CID | 24085 |
ChEBI | CHEBI:31823 |
SMILES | [Cl-].[Cl-].[Hg++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316927
|
Thermo Scientific Alfa Aesar
010808.14 |
25 g |
78.89 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306927
|
Thermo Scientific Alfa Aesar
010808.22 |
100 g |
379.89 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326927
|
Thermo Scientific Alfa Aesar
010808.36 |
500 g | N/A | ||||||
Description
Mercuric chloride is used as a catalyst for the conversion of acetylene to vinyl chloride, the precursor to polyvinylchloride It is used as embalming chemical, photographic intensifier. It is also used as a treatment for syphilis and in very dilute solution as an antiseptic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7487-94-7 | |
271.49 | |
LWJROJCJINYWOX-UHFFFAOYSA-L | |
24085 | |
[Cl-].[Cl-].[Hg++] |
Cl2Hg | |
MFCD00011041 | |
mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 | |
CHEBI:31823 |
Specifications
302°C | |
7487-94-7 | |
Powder | |
HgCl2 | |
UN1624 | |
mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2 | |
[Cl-].[Cl-].[Hg++] | |
271.49 | |
CHEBI:31823 | |
99.999% | |
Odorless | |
5.4g/mL | |
Mercury(II) chloride |
280°C to 282°C | |
White | |
Cl2Hg | |
MFCD00011041 | |
14,5876 | |
LWJROJCJINYWOX-UHFFFAOYSA-L | |
dichloromercury | |
24085 | |
271.5 | |
Puratronic™ | |
1.8590 | |
(metals basis) |
Safety and Handling
GHS H Statement
H300-H372-H341-H361-H314
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURIC CHLORIDE
missing translation for 'rtecsNumber' : OV9100000
missing translation for 'tsca' : TSCA
RUO – Research Use Only