missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11306927
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Mercuric chloride is used as a catalyst for the conversion of acetylene to vinyl chloride, the precursor to polyvinylchloride It is used as embalming chemical, photographic intensifier. It is also used as a treatment for syphilis and in very dilute solution as an antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-94-7
Molecular Formula Cl2Hg
Molecular Weight (g/mol) 271.49
MDL Number MFCD00011041
InChI Key LWJROJCJINYWOX-UHFFFAOYSA-L
Synonym mercuric chloride, mercury ii chloride, sublimate, mercury bichloride, mercury dichloride, corrosive sublimate, mercuric bichloride, mercury perchloride, sublimat, hgcl2
PubChem CID 24085
ChEBI CHEBI:31823
SMILES [Cl-].[Cl-].[Hg++]

Specifications

Boiling Point 302°C
Melting Point 280°C to 282°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula HgCl2
UN Number UN1624
Merck Index 14,5876
Solubility Information Soluble in water,alcohol,acetone and ethyl acetate.
IUPAC Name mercury(2+) dichloride
Formula Weight 271.5
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Refractive Index 1.859
Density 5.4 g/mL
Assay (metals basis)
Chemical Name or Material Mercury(II) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'