Learn More
Masterflex™ Tygon™ E-LFL Transparent Pump Tubing
Use this high-performance pump tubing for challenging applications. Masterflex™ Transparent Tygon™ E-LFL Pump Tubing won’t age or oxidize, fits tightly to glass and metal and bends to small radius curves.
118.00 € - 740.00 €
Specifications
Autoclavable Autoclavable
Material Tygon E-LFL
Max. Operating Temperature (Metric) 74°C
View More Specs
Products 13
Product Code Brand Tubing Size Product Type Price Quantity & Availability  
Product Code Brand Tubing Size Product Type Price Quantity & Availability  
12863383
View Documents
Masterflex™
06440-13
13 L/S Pump Tubing
118.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12833393
View Documents
Masterflex™
06440-26
26 I/P Pump Tubing
325.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12853383
View Documents
Masterflex™
06440-14
14 L/S Pump Tubing
145.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12863393
View Documents
Masterflex™
06440-70
70 I/P Pump Tubing
575.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12893383
View Documents
Masterflex™
06440-15
15 L/S Pump Tubing
183.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12883383
View Documents
Masterflex™
06440-17
17 L/S Pump Tubing
187.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12803393
View Documents
Masterflex™
06440-18
18 L/S Pump Tubing
211.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12823393
View Documents
Masterflex™
06440-25
25 L/S Pump Tubing
163.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12843393
View Documents
Masterflex™
06440-35
35 L/S Pump Tubing
271.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12873393
View Documents
Masterflex™
06440-73
73 I/P Pump Tubing
312.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12883393
View Documents
Masterflex™
06440-82
82 I/P Pump Tubing
407.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12823403
View Documents
Masterflex™
06440-88
88 I/P Pump Tubing
710.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
12813403
View Documents
Masterflex™
06440-89
89 I/P Pump Tubing
740.00 €
1 roll
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
Use this high-performance pump tubing for challenging applications. Masterflex™ Transparent Tygon™ E-LFL Pump Tubing won’t age or oxidize, fits tightly to glass and metal and bends to small radius curves.
  • Exceeds USP class VI, 3A sanitary standards
  • FDA compliant
  • Nontoxic and non-hemolytic with excellent biocompatibility
  • Longest life of all Tygon formulations
  • Pump at continuous pressures up to 40psi
  • Low extractables and low gas permeability
Specifications
Specifications
Autoclavable
Tygon E-LFL
74°C