Learn More
Lithium ethoxide, 1M solution in ethanol, Thermo Scientific Chemicals
83.00 € - 393.00 €
Chemical Identifiers
CAS | 2388-07-0, 64-17-5 |
---|---|
Molecular Formula | C2H5LiO |
Molecular Weight (g/mol) | 52.00 |
MDL Number | MFCD00050493 |
InChI Key | AZVCGYPLLBEUNV-UHFFFAOYSA-N |
Synonym | lithium ethoxide, lithium ethylate, lithotab ethoxide, lioet, acmc-1cex9, ethanol, lithium salt 9ci, lithium 1+ ion ethoxide |
PubChem CID | 23661847 |
SMILES | [Li+].CC[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10536843
|
Thermo Scientific Acros
380171000 |
100 mL | AcroSeal™ Glass Bottle |
83.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10298983
|
Thermo Scientific Acros
380178000 |
800 mL | AcroSeal™ Glass Bottle |
393.00 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2388-07-0, 64-17-5 | |
52.00 | |
AZVCGYPLLBEUNV-UHFFFAOYSA-N | |
23661847 |
C2H5LiO | |
MFCD00050493 | |
lithium ethoxide, lithium ethylate, lithotab ethoxide, lioet, acmc-1cex9, ethanol, lithium salt 9ci, lithium 1+ ion ethoxide | |
[Li+].CC[O-] |
Specifications
Orange | |
C2H5LiO | |
MFCD00050493 | |
0.8200g/mL | |
AZVCGYPLLBEUNV-UHFFFAOYSA-N | |
lithium;ethanolate | |
23661847 | |
Confirmed | |
Lithium ethoxide, 1M solution in ethanol |
2388-07-0, 64-17-5 | |
LiOCH2CH3 | |
Exact strength on the certificate of analysis | |
lithium ethoxide, lithium ethylate, lithotab ethoxide, lioet, acmc-1cex9, ethanol, lithium salt 9ci, lithium 1+ ion ethoxide | |
[Li+].CC[O-] | |
52.00 | |
52 | |
Liquid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Reacts violently with water.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
For Manufacturing and Laboratory Use. Not for beverage use