missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium ethoxide, 1M solution in ethanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Orange |
Linear Formula | LiOCH2CH3 |
Concentration | Exact strength on the certificate of analysis |
Density | 0.8200g/mL |
IUPAC Name | lithium(1+) ethanolate |
Formula Weight | 52 |
Identification | Confirmed |
Packaging | AcroSeal™ Glass Bottle |
Quantity | 800 mL |
Physical Form | Liquid |
Show More |
For Manufacturing and Laboratory Use. Not for beverage use
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'