missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Chloride, ReagentPlus™, Honeywell™
Reagent Grade, 99%
60.60 € - 494.62 €
Chemical Identifiers
CAS | 7447-41-8 |
---|---|
Molecular Formula | ClLi |
Molecular Weight (g/mol) | 42.39 |
MDL Number | MFCD00011078 |
InChI Key | KWGKDLIKAYFUFQ-UHFFFAOYSA-M |
Synonym | lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride |
PubChem CID | 433294 |
ChEBI | CHEBI:48607 |
IUPAC Name | lithium;chloride |
SMILES | [Li+].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15601960
|
Honeywell Research Chemicals
213233-100G |
100 g |
60.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15621960
|
Honeywell Research Chemicals
213233-500G |
500 g | |||||||
15611960
|
Honeywell Research Chemicals
213233-2KG |
2 kg |
494.62 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7447-41-8 | |
42.39 | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
433294 | |
lithium;chloride |
ClLi | |
MFCD00011078 | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
CHEBI:48607 | |
[Li+].[Cl-] |
Specifications
7447-41-8 | |
LiCl | |
NONH for all modes of transport | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
lithium;chloride | |
433294 | |
42.39g/mol | |
ReagentPlus™ |
ClLi | |
MFCD00011078 | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
[Li+].[Cl-] | |
42.39 | |
CHEBI:48607 | |
99% | |
Lithium Chloride |
Safety and Handling
P301 + P312 + P330-P305 + P351 + P338
H302-H315-H319
missing translation for 'einecsNumber' : 231-212-3
missing translation for 'rtecsNumber' : OJ5950000