missing translation for 'onlineSavingsMsg'
Learn More

Lithium Chloride, ReagentPlus™, Honeywell™

Product Code. 15621960
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium;chloride
SMILES [Li+].[Cl-]

Specifications

Quantity 500 g
Linear Formula LiCl
UN Number NONH for all modes of transport
Formula Weight 42.39g/mol
Percent Purity 99%
Grade ReagentPlus™
Chemical Name or Material Lithium Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.