missing translation for 'onlineSavingsMsg'
Learn More

Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole, 95%, Thermo Scientific Chemicals

61.71 € - 369.84 €

Chemical Identifiers

CAS 761441-54-7
Molecular Formula C6F3LiN4
Molecular Weight (g/mol) 192.03
MDL Number MFCD18827480
InChI Key CVVIFWCYVZRQIY-UHFFFAOYSA-N
Synonym litdi, lithium 4,5-dicyano-2-trifluoromethyl imidazole, lithium 4,5-dicyano-2-trifluoromethyl-imidazolide, 1-lithio-2-trifluoromethyl-1h-imidazole-4,5-dicarbonitrile
PubChem CID 73995168
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15438775
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H34081.03
1 g
61.71 €
1g
Please to purchase this item. Need a web account? Register with us today!
15448775
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H34081.09
10 g
369.84 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

761441-54-7
192.03
CVVIFWCYVZRQIY-UHFFFAOYSA-N
73995168
C6F3LiN4
MFCD18827480
litdi, lithium 4,5-dicyano-2-trifluoromethyl imidazole, lithium 4,5-dicyano-2-trifluoromethyl-imidazolide, 1-lithio-2-trifluoromethyl-1h-imidazole-4,5-dicarbonitrile

Specifications

761441-54-7
C6F3LiN4
litdi, lithium 4,5-dicyano-2-trifluoromethyl imidazole, lithium 4,5-dicyano-2-trifluoromethyl-imidazolide, 1-lithio-2-trifluoromethyl-1h-imidazole-4,5-dicarbonitrile
192.03
192.03
Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole
>300°C
MFCD18827480
CVVIFWCYVZRQIY-UHFFFAOYSA-N
73995168
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 691-963-1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only