missing translation for 'onlineSavingsMsg'
Learn More

Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole, 95%, Thermo Scientific Chemicals

Product Code. 15448775
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 761441-54-7
Molecular Formula C6F3LiN4
Molecular Weight (g/mol) 192.03
MDL Number MFCD18827480
InChI Key CVVIFWCYVZRQIY-UHFFFAOYSA-N
Synonym litdi, lithium 4,5-dicyano-2-trifluoromethyl imidazole, lithium 4,5-dicyano-2-trifluoromethyl-imidazolide, 1-lithio-2-trifluoromethyl-1h-imidazole-4,5-dicarbonitrile
PubChem CID 73995168

Specifications

Melting Point >300°C
Quantity 10 g
Formula Weight 192.03
Percent Purity 95%
Chemical Name or Material Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'