missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) carbonate, ACS, Thermo Scientific Chemicals

Maximum level of impurities: Insoluble in dilute acetic acid 0.02%, Cl 0.002%, Nitrate and nitrite (as NO3) P.T. (limit about 0.005%), Ca 0.01%, K 0.02%, Na 0.05%, Cd 0.002%, Fe 0.005%, Zn 0.003%

71.25 € - 712.55 €

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
MDL Number MFCD00064631
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
Synonym lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
PubChem CID 11727
SMILES [Pb++].[O-]C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11319257
View Documents
Thermo Scientific Alfa Aesar
040119.22
100 g
71.25 €
100g
Please to purchase this item. Need a web account? Register with us today!
11309257
View Documents
Thermo Scientific Alfa Aesar
040119.36
500 g
294.47 €
500g
Please to purchase this item. Need a web account? Register with us today!
11399247
View Documents
Thermo Scientific Alfa Aesar
040119.57
4 x 500 g
712.55 €
2000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lead(II) carbonate is used in polymerization catalysts. It serves as a curing agent for silicon resins, heat-resistant lubricants, lead-acid storage batteries, wire-reinforced hoses, thermistors and heat stabilizers for polyvinyl chloride polymers. Further, it is used in electrophotography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

598-63-0
267.20
MFEVGQHCNVXMER-UHFFFAOYSA-L
11727
CO3Pb
MFCD00064631
lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
[Pb++].[O-]C([O-])=O

Specifications

598-63-0
Powder
MFCD00064631
Soluble in acid and alkali. Insoluble in water,alcohol and ammonia.
[Pb++].[O-]C([O-])=O
267.20
267.2
Odorless
6.6g/cm3 at 20°C
White
CO3Pb
lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
MFEVGQHCNVXMER-UHFFFAOYSA-L
lead(2+);carbonate
11727
ACS
White powder
Lead(II) carbonate, For analysis ACS
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

missing translation for 'dotInformation' : DOT Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 209-943-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Lead(II) carbonate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.