missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) carbonate, ACS, Thermo Scientific Chemicals

Product Code. 11309257
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Maximum level of impurities: Insoluble in dilute acetic acid 0.02%, Cl 0.002%, Nitrate and nitrite (as NO3) P.T. (limit about 0.005%), Ca 0.01%, K 0.02%, Na 0.05%, Cd 0.002%, Fe 0.005%, Zn 0.003%

Lead(II) carbonate is used in polymerization catalysts. It serves as a curing agent for silicon resins, heat-resistant lubricants, lead-acid storage batteries, wire-reinforced hoses, thermistors and heat stabilizers for polyvinyl chloride polymers. Further, it is used in electrophotography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-63-0
Molecular Formula CO3Pb
Molecular Weight (g/mol) 267.20
MDL Number MFCD00064631
InChI Key MFEVGQHCNVXMER-UHFFFAOYSA-L
Synonym lead carbonate, lead ii carbonate, cerussete, cerussite, lead 2+ carbonate, plumbous carbonate, dibasic lead carbonate, unii-43m0p24l2b, carbonic acid, lead 2+ salt 1:1, lead carbonate pbco3
PubChem CID 11727
SMILES [Pb++].[O-]C([O-])=O

Specifications

Physical Form Powder
Quantity 500 g
UN Number UN2291
Solubility Information Soluble in acid and alkali. Insoluble in water,alcohol and ammonia.
IUPAC Name λ²-lead(2+) carbonate
Formula Weight 267.2
Grade ACS
Odor Odorless
Chemical Name or Material Lead(II) carbonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.