missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Dispensing Safety Cans
Reliable dispensing features at an economical price
307.00 € - 325.75 €
Specifications
Certifications/Compliance | FM, CARB, NFPA, OSHA |
---|---|
Closure Type | Self Closing Pressure Relief Cap |
Capacity (English) | 5 gal. |
Capacity (Metric) | 19 L |
Color | Red |
Product Code | Brand | Product Type | Height (English) | Height (Metric) | Includes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Height (English) | Height (Metric) | Includes | Price | Quantity & Availability | ||
12981385
|
Justrite™
7150140 |
Type I Shelf Safety Can | 16.8 in. | 429 mm | Tap 08540 |
307.00 €
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||
12911395
|
Justrite™
7150156 |
Type I Tilt Safety Can | 15.8 in. | 403 mm | Tap 08902 and Stand | ||||
Description
For safely dispensing hazardous liquids into test tubes, small flasks and beakers
- Vacuum-relief valve within the lid for smooth liquid flow and self-closing faucets for trouble-free dispensing
- Fill spouts have self-close, pressure-relief caps and stainless steel flame arresters
- Capacity: 19L (5 gal)
- Color: Red
- Smaller 19-413-847 faucet offers easier dispensing in lab environments; No. 08902 faucet better suited for heavy, industrial use
Shelf Cans
- Faucet mounted two inches above the bottom to eliminate lifting the cans during dispensing
- Faucet on top with a sturdy metal cradle for the can; cradle base measures 35.1 × 35.6cm (13.81 × 14 in.)
Certifications
FM
Compliance
CARB, NFPA, OSHA
Specifications
FM, CARB, NFPA, OSHA | |
5 gal. | |
Red | |
Steel | |
1.58 in. | |
4 cm |
Self Closing Pressure Relief Cap | |
19 L | |
For Laboratories | |
11.75 in. | |
298 mm |