missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ Jackson Safety™ G80 Chemical Resistant Nitrile Gloves
Green, hand-specific gloves providing maximum protection against chemical and mechanical hazards. KleenGuard™ Jackson Safety™ G80 Chemical Resistant Nitrile Gloves suitable for various applications.
138.00 €
Specifications
Color | Green |
---|---|
Finger Thickness (English) | 22 mil |
Finger Thickness (Metric) | 0.558 mm |
Latex Free | Yes |
Material | Nitrile |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15376728
|
Kimberly-Clark Professional™
25622 |
8 |
138.00 €
12 pairs |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
15386728
|
Kimberly-Clark Professional™
25623 |
9 | |||||||
15306738
|
Kimberly-Clark Professional™
25624 |
10 | |||||||
Description
- Suitable for various applications and offer reliable protection against a wide range of chemicals and also mechanical hazards
- Ideal for: providing PPE Category 3 protection in many different segments like Aerospace, Automotive, Metal Manufacturing, Food, Transportation, Construction, when handling chemicals, oils, grease, alcohols, acids, bases or solvents also in immersion and providing the highest level of abrasion resistance
- Risk of linting is avoided protecting the products from contamination
- Longer length to protect wrist and lower forearm and allow full immersion
- With sand patch texturing on fingers and palm offering enhanced wet and dry grip
- AQL 0.65 for pinholes to provide superior protection, more productivity and efficiency
Specifications
Green | |
0.558 mm | |
Nitrile | |
0.55 mm | |
21.7 mil | |
47.8 x 15.6 cm | |
47.8 cm |
22 mil | |
Yes | |
21.7 mil | |
Chemical-resistant Gloves | |
0.5 mm | |
18.642 in. | |
45 cm |