missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ Jackson Safety™ G80 Chemical Resistant Nitrile Gloves

Product Code. 15376728
Click to view available options
Size:
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Green, hand-specific gloves providing maximum protection against chemical and mechanical hazards. KleenGuard™ Jackson Safety™ G80 Chemical Resistant Nitrile Gloves suitable for various applications.

  • Suitable for various applications and offer reliable protection against a wide range of chemicals and also mechanical hazards
  • Ideal for: providing PPE Category 3 protection in many different segments like Aerospace, Automotive, Metal Manufacturing, Food, Transportation, Construction, when handling chemicals, oils, grease, alcohols, acids, bases or solvents also in immersion and providing the highest level of abrasion resistance
  • Risk of linting is avoided protecting the products from contamination
  • Longer length to protect wrist and lower forearm and allow full immersion
  • With sand patch texturing on fingers and palm offering enhanced wet and dry grip
  • AQL 0.65 for pinholes to provide superior protection, more productivity and efficiency

Specifications

Color Green
Finger Thickness (English) 22 mil
Finger Thickness (Metric) 0.558 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 21.7 mil
Palm Thickness (Metric) 0.55 mm
Product Type Chemical-resistant Gloves
Size 8
Thickness (English) 21.7 mil
Thickness (Metric) 0.5 mm
Dimensions (L x W) 47.8 x 15.6 cm
Length (English) 18.642 in.
Length (Metric) 47.8 cm
Length (Metric) Cuff 45 cm
Weight (English) 4.0656 lb.
Weight (Metric) 1.848 kg
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'