missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-103.4% (manganometric), Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-103.4% (manganometric)

23.33 € - 638.10 €

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name iron(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15643310
View Documents Promotion Details
Honeywell-Fluka
31236-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15673310
View Documents Promotion Details
Honeywell-Fluka
31236-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15653310
View Documents Promotion Details
Honeywell-Fluka
31236-1KG
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15663310
View Documents Promotion Details
Honeywell-Fluka
31236-25KG
25 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7782-63-0
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
62662
iron(2+);sulfate;heptahydrate
FeH14O11S
MFCD00149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
CHEBI:75836
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

7782-63-0
FeSO4 · 7H2O
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
278.01
CHEBI:75836
99 to 103.4% (manganometric)
14.6mmHg (25°C)
Iron(Ii) Sulfate Heptahydrate, Reag. Ph. Eur. reag. ISO
FeH14O11S
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron(2+);sulfate;heptahydrate
62662
278.01g/mol
ACS/puriss. p.a.
1.898g/mL at 25°C
Videos
Safety and Handling

Safety and Handling

P301 + P312 + P330-P305 + P351 + P338

H302-H315-H319

missing translation for 'einecsNumber' : 231-753-5

missing translation for 'rtecsNumber' : NO8510000

Product Identifier
  • Iron(II) sulfate heptahydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : iron(II)sulphate-7-hydrate

SDS
Documents

Documents

Product Certifications