missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-103.4% (manganometric), Honeywell Fluka™

Product Code. 15663310
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-103.4% (manganometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name iron(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100.0000%
Quantity 25 kg
Assay Percent Range 99.0 to 103.4%
Linear Formula NH4VO3
Formula Weight 278.01
Percent Purity 99.0 to 103.4% (manganometric)
Grade ACS Reagent/Reagent ISO/Reagent Ph. Eur.
Packaging Paper / Poly Bag
Vapor Pressure 14.6mmHg (25°C)
Density 1.898 g/mL at 25 °C(lit.)
Chemical Name or Material Iron(II) sulfate heptahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.0-103.4% (manganometric)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'